siRNA Details
| Gene Symbol: RPS24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001026.4 | NP_001017.1 | 3 | 264 | ||
| NM_001142282.1 | NP_001135754.1 | 3 | 264 | |||
| NM_001142283.1 | NP_001135755.1 | 3 | 264 | |||
| NM_001142284.1 | NP_001135756.1 | 3 | 264 | |||
| NM_001142285.1 | NP_001135757.1 | 3 | 264 | |||
| NM_033022.3 | NP_148982.1 | 3 | 264 | |||
| GenBank mRNA | AF147330.1 | - | 3 | 145 | ||
| AK311776.1 | - | 3 | 161 | |||
| AK311910.1 | - | 3 | 161 | |||
| BC000523.2 | - | 3 | 141 | 2961125 | ||
| BC003149.1 | - | 3 | 137 | |||
| BC071926.1 | - | 3 | 146 | 5556377 | ||
| BG610671.1 | - | 208 | ||||
| BG944046.1 | - | 3 | 265 | |||
| BM745909.1 | - | 3 | 161 | |||
| BX537975.1 | - | 3 | 160 | |||
| BX647260.1 | - | 3 | 150 | |||
| CD176901.1 | - | 3 | 187 | |||
| CD677556.1 | - | 3 | 140 | |||
| DB116225.1 | - | 3 | 161 | |||
| M31520.1 | - | 3 | 264 | |||

RPS24
Genomic Map