siRNA Details
| Gene Symbol: WDR62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083961.1 | NP_001077430.1 | 15 | 1945 | ||
| NM_173636.4 | NP_775907.4 | 15 | 1945 | |||
| XM_005258809.2 | XP_005258866.1 | 15 | 1945 | |||
| XM_011526837.1 | XP_011525139.1 | 15 | 1930 | |||
| XM_011526838.1 | XP_011525140.1 | 15 | 1945 | |||
| XM_011526839.1 | XP_011525141.1 | 13 | 1594 | |||
| XM_011526840.2 | XP_011525142.1 | 8 | 940 | |||
| XM_011526841.2 | XP_011525143.1 | 6 | 561 | |||
| XM_011526842.1 | XP_011525144.1 | 5 | 393 | |||
| XM_017026665.1 | XP_016882154.1 | 15 | 1945 | |||
| GenBank mRNA | BC058939.1 | - | 15 | 1889 | ||
| BQ880028.1 | - | 2 | 114 | |||
| BQ960310.1 | - | 5 | 442 | |||
| BX647726.1 | - | 16 | 2006 | |||

WDR62
Genomic Map