siRNA Details
| Gene Symbol: GNPTAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024312.4 | NP_077288.2 | 13, 14 | 2916 | ||
| XM_006719593.2 | XP_006719656.1 | 13, 14 | 2911 | |||
| XM_011538731.2 | XP_011537033.1 | 13, 14 | 2912 | |||
| XM_017019961.1 | XP_016875450.1 | 13, 14 | 2813 | |||
| XM_017019962.1 | XP_016875451.1 | 13, 14 | 3018 | |||
| GenBank mRNA | AB033034.2 | - | 6, 7 | 1807 | ||
| AM085438.1 | - | 13, 14 | 2903 | |||
| AY687932.1 | - | 13, 14 | 2888 | |||
| BC042615.1 | - | 1, 2 | 427 | |||
| BC131787.1 | - | 13, 14 | 2844 | 40126580 | ||

GNPTAB
Genomic Map