siRNA Details
| Gene Symbol: LRIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198506.4 | NP_940908.3 | 4 | 1443 | ||
| XM_017008167.1 | XP_016863656.1 | 3 | 1065 | |||
| GenBank mRNA | AK126648.1 | - | 4 | 1640 | ||
| BC104037.2 | - | 3 | 1104 | 40026462 | ||

LRIT3
Genomic Map