siRNA Details
| Gene Symbol: FLJ22763 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033977.1 | 3 | 1877 | |||
| GenBank mRNA | AK026416.1 | - | 3 | 1873 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ22763| Gene Symbol: FLJ22763 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033977.1 | 3 | 1877 | |||
| GenBank mRNA | AK026416.1 | - | 3 | 1873 | ||