siRNA Details
| Gene Symbol: TCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000355.3 | NP_000346.2 | 5, 6 | 1002 | ||
| NM_001184726.1 | NP_001171655.1 | 5, 6 | 921 | |||
| GenBank mRNA | AK225445.1 | - | 5, 6 | 791 | ||
| AK313711.1 | - | 5, 6 | 892 | |||
| BC001176.1 | - | 5, 6 | 880 | 3353271 | ||
| BC011239.1 | - | 5, 6 | 800 | 4153362 | ||
| CR456591.1 | - | 5, 6 | 792 | |||
| L02647.1 | - | 5, 6 | 850 | |||
| L02648.1 | - | 5, 6 | 806 | |||
| M60396.1 | - | 5, 6 | 790 | |||

TCN2
Genomic Map