siRNA Details
| Gene Symbol: GCDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000159.3 | NP_000150.1 | 6 | 547 | ||
| NM_013976.3 | NP_039663.1 | 6 | 547 | |||
| NR_102316.1 | 6 | 602 | ||||
| NR_102317.1 | 5 | 855 | ||||
| XM_006722721.2 | XP_006722784.1 | 6 | 516 | |||
| XM_011527899.2 | XP_011526201.1 | 6 | 516 | |||
| XM_011527900.2 | XP_011526202.1 | 6 | 516 | |||
| XM_017026580.1 | XP_016882069.1 | 6 | 516 | |||
| GenBank mRNA | AK098370.1 | - | 6 | 516 | ||
| AK290407.1 | - | 6 | 516 | |||
| AK296446.1 | - | 6 | 565 | |||
| AK298772.1 | - | 4 | 417 | |||
| AK300841.1 | - | 5 | 804 | |||
| AK309892.1 | - | 5 | 804 | |||
| AU135567.1 | - | 6 | 516 | |||
| BC002579.2 | - | 6 | 500 | 3142978 | ||
| BT006706.1 | - | 5 | 439 | |||
| U69141.1 | - | 5 | 475 | |||

GCDH
Genomic Map