siRNA Details
| Gene Symbol: GCDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000159.3 | NP_000150.1 | 12 | 1670 | ||
| NM_013976.3 | NP_039663.1 | 12 | 1438 | |||
| NR_102316.1 | 12 | 1725 | ||||
| NR_102317.1 | 11 | 1943 | ||||
| GenBank mRNA | AK098370.1 | - | 12 | 1638 | ||
| AK290407.1 | - | 12 | 1639 | |||
| AK296446.1 | - | 12 | 1688 | |||
| BC002579.2 | - | 12 | 1623 | 3142978 | ||
| BM855466.1 | - | 1 | 281 | |||
| U69141.1 | - | 11 | 1598 | |||

GCDH
Genomic Map