siRNA Details
| Gene Symbol: GCDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006722721.2 | XP_006722784.1 | 12 | 1337 | ||
| XM_011527900.2 | XP_011526202.1 | 12 | 1337 | |||
| XM_017026580.1 | XP_016882069.1 | 12 | 1337 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GCDH| Gene Symbol: GCDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006722721.2 | XP_006722784.1 | 12 | 1337 | ||
| XM_011527900.2 | XP_011526202.1 | 12 | 1337 | |||
| XM_017026580.1 | XP_016882069.1 | 12 | 1337 | |||