siRNA Details
| Gene Symbol: LZTFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276378.1 | NP_001263307.1 | 12 | 2348 | ||
| NM_001276379.1 | NP_001263308.1 | 10 | 2119 | |||
| NM_020347.3 | NP_065080.1 | 10 | 2067 | |||
| NR_075080.1 | 9 | 1942 | ||||
| XM_006713207.2 | XP_006713270.2 | 9 | 1948 | |||
| XM_011533838.2 | XP_011532140.1 | 10 | 1991 | |||
| XM_017006644.1 | XP_016862133.1 | 13 | 2705 | |||
| XM_017006645.1 | XP_016862134.1 | 12 | 2580 | |||
| GenBank mRNA | AJ297351.1 | - | 10 | 2024 | ||
| AK027165.1 | - | 1 | 695 | |||
| AK129805.1 | - | 1 | 111 | |||
| BC042483.1 | - | 10 | 2027 | 4831113 | ||
| BX640604.1 | - | 10 | 2049 | |||

LZTFL1
Genomic Map