siRNA Details
| Gene Symbol: A4GALT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318038.1 | NP_001304967.1 | 3 | 1272 | ||
| NM_017436.5 | NP_059132.1 | 3 | 1249 | |||
| XM_005261644.2 | XP_005261701.1 | 3 | 1554 | |||
| XM_005261646.4 | XP_005261703.1 | 2 | 1110 | |||
| XM_005261647.2 | XP_005261704.1 | 3 | 1084 | |||
| XM_005261648.3 | XP_005261705.1 | 3 | 1167 | |||
| XM_006724265.3 | XP_006724328.1 | 3 | 1495 | |||
| XM_006724266.3 | XP_006724329.1 | 3 | 1229 | |||
| XM_011530233.2 | XP_011528535.1 | 2 | 1436 | |||
| XM_017028831.1 | XP_016884320.1 | 2 | 1366 | |||
| GenBank mRNA | AB037883.1 | - | 2 | 1081 | ||
| AJ245581.1 | - | 1 | 948 | |||
| AK074388.1 | - | 1 | 848 | |||
| AK312927.1 | - | 3 | 1253 | |||
| BC017068.2 | - | 3 | 1168 | 3913851 | ||
| BC055286.1 | - | 2 | 1438 | 6161307 | ||
| KU178649.1 | - | 1 | 948 | |||
| KU178650.1 | - | 2 | 567 | |||

A4GALT
Genomic Map