siRNA Details
| Gene Symbol: AMTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286731.1 | NP_001273660.1 | 5 | 366 | ||
| NM_212557.3 | NP_997722.1 | 5 | 369 | |||
| GenBank mRNA | AB593161.1 | - | 5 | 368 | ||
| AY358528.1 | - | 5 | 360 | |||
| BC121812.1 | - | 5 | 337 | 40109020 | ||
| BC121817.1 | - | 5 | 340 | 40109016 | ||

AMTN
Genomic Map