siRNA Details
| Gene Symbol: CEP57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243776.1 | NP_001230705.1 | 12 | 1752 | ||
| NM_001243777.1 | NP_001230706.1 | 10 | 1630 | |||
| NM_014679.4 | NP_055494.2 | 11 | 1708 | |||
| XM_006718945.2 | XP_006719008.1 | 10 | 1591 | |||
| XM_006718946.2 | XP_006719009.1 | 9 | 1522 | |||
| XM_017018592.1 | XP_016874081.1 | 12 | 2898 | |||
| XM_017018593.1 | XP_016874082.1 | 9 | 1513 | |||
| XM_017018594.1 | XP_016874083.1 | 8 | 1405 | |||
| GenBank mRNA | AK098647.1 | - | 11 | 1681 | ||
| AK291245.1 | - | 11 | 1663 | |||
| AK293277.1 | - | 12 | 1705 | |||
| AY225092.1 | - | 11 | 1470 | |||
| AY239292.1 | - | 11 | 1609 | |||
| BC001233.2 | - | 11 | 1691 | 3451715 | ||
| BC031048.1 | - | 11 | 1619 | |||
| D42054.1 | - | 10 | 1445 | |||

CEP57
Genomic Map