siRNA Details
| Gene Symbol: AMTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286731.1 | NP_001273660.1 | 6, 7 | 435 | ||
| NM_212557.3 | NP_997722.1 | 6, 7 | 438 | |||
| GenBank mRNA | AB593161.1 | - | 6, 7 | 437 | ||
| AY358528.1 | - | 6, 7 | 429 | |||
| BC121812.1 | - | 6, 7 | 406 | 40109020 | ||
| BC121817.1 | - | 6, 7 | 409 | 40109016 | ||

AMTN
Genomic Map