siRNA Details
| Gene Symbol: MMP26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021801.4 | NP_068573.2 | 2, 3 | 349 | ||
| XM_011520219.2 | XP_011518521.1 | 3, 4 | 529 | |||
| GenBank mRNA | AF230354.1 | - | 2, 3 | 333 | ||
| AF248646.1 | - | 2, 3 | 337 | |||
| AF291664.1 | - | 2, 3 | 346 | |||
| AJ251531.1 | - | 2, 3 | 343 | |||
| BC101541.1 | - | 2, 3 | 349 | 8069047 | ||
| BC101543.1 | - | 2, 3 | 349 | 8069049 | ||

MMP26
Genomic Map