siRNA Details
| Gene Symbol: HOXA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006735.3 | NP_006726.1 | 1 | 368 | ||
| GenBank mRNA | AK096742.1 | - | 1 | 279 | ||
| AK291164.1 | - | 1 | 277 | |||
| BC130571.1 | - | 1 | 122 | 40146602 | ||
| BC136500.1 | - | 1 | 122 | 9020489 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: BC027688, AB221621, BC117107, BC117105, M95599, and others.

HOXA2
Genomic Map