siRNA Details
| Gene Symbol: PMS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000535.6 | NP_000526.2 | 10 | 1117 | ||
| NM_001322003.1 | NP_001308932.1 | 10 | 1112 | |||
| NM_001322004.1 | NP_001308933.1 | 9 | 977 | |||
| NM_001322005.1 | NP_001308934.1 | 10 | 1229 | |||
| NM_001322007.1 | NP_001308936.1 | 9 | 1014 | |||
| NM_001322008.1 | NP_001308937.1 | 8 | 874 | |||
| NM_001322009.1 | NP_001308938.1 | 10 | 1224 | |||
| NM_001322011.1 | NP_001308940.1 | 10 | 1068 | |||
| NM_001322012.1 | NP_001308941.1 | 10 | 1063 | |||
| NM_001322013.1 | NP_001308942.1 | 9 | 944 | |||
| NM_001322014.1 | NP_001308943.1 | 10 | 1117 | |||
| NM_001322015.1 | NP_001308944.1 | 10 | 1287 | |||
| NR_136154.1 | 10 | 1117 | ||||
| XM_006715744.3 | XP_006715807.1 | 3 | 193 | |||
| XM_017012342.1 | XP_016867831.1 | 9 | 942 | |||
| GenBank mRNA | AB103082.1 | - | 10 | 1030 | ||
| AB103086.1 | - | 7 | 712 | |||
| AK294661.1 | - | 9 | 967 | |||
| AK312390.1 | - | 10 | 1066 | |||
| BC008400.1 | - | 10 | 1075 | |||
| BC031832.1 | - | 10 | 1081 | |||
| BC093921.1 | - | 10 | 1052 | 7939766 | ||
| U14658.1 | - | 10 | 1032 | |||

PMS2
Genomic Map