siRNA Details
| Gene Symbol: PRX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020956.2 | NP_066007.1 | 6 | 3076 | ||
| NM_181882.2 | NP_870998.2 | 7 | 2427 | |||
| XM_011527171.2 | XP_011525473.1 | 7 | 2419 | |||
| XM_017027046.1 | XP_016882535.1 | 3 | 2578 | |||
| XM_017027047.1 | XP_016882536.1 | 4 | 2121 | |||
| GenBank mRNA | AB046840.1 | - | 3 | 2557 | ||
| AF321191.1 | - | 7 | 2427 | |||
| AF321192.1 | - | 6 | 3076 | |||
| BC067266.1 | - | 7 | 2429 | 6177899 | ||

PRX
Genomic Map