siRNA Details
| Gene Symbol: TSPAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004615.3 | NP_004606.2 | 1 | 124 | ||
| GenBank mRNA | AB062057.1 | - | 1 | 125 | ||
| AB102665.1 | - | 1 | 40 | |||
| AK293178.1 | - | 1 | 58 | |||
| AK293534.1 | - | 1 | 60 | |||
| AK293552.1 | - | 1 | 58 | |||
| AK312343.1 | - | 1 | 58 | |||
| BC018036.1 | - | 1 | 83 | 4801388 | ||
| D10653.1 | - | 1 | 64 | |||
| D29808.1 | - | 1 | 50 | |||
| DB475293.1 | - | 1 | 233 | |||
| L10373.1 | - | 1 | 108 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: AK004776, BC058603, BY238972, BC050153, AK147404, and others.

TSPAN7
Genomic Map