siRNA Details
| Gene Symbol: TSPAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004615.3 | NP_004606.2 | 6 | 701 | ||
| GenBank mRNA | AB062057.1 | - | 6 | 702 | ||
| AB102665.1 | - | 6 | 617 | |||
| AK293178.1 | - | 7 | 686 | |||
| AK293534.1 | - | 8 | 855 | |||
| AK293552.1 | - | 7 | 766 | |||
| AK312343.1 | - | 6 | 635 | |||
| BC018036.1 | - | 6 | 660 | 4801388 | ||
| D10653.1 | - | 6 | 641 | |||
| D29808.1 | - | 6 | 627 | |||
| L10373.1 | - | 6 | 685 | |||

TSPAN7
Genomic Map