siRNA Details
| Gene Symbol: FLJ33360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028351.1 | 2 | 252 | |||
| GenBank mRNA | AK090679.1 | - | 2 | 252 | ||
| BC132707.1 | - | 2 | 240 | 40146729 | ||
| BC132709.1 | - | 2 | 240 | 40146731 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ33360| Gene Symbol: FLJ33360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028351.1 | 2 | 252 | |||
| GenBank mRNA | AK090679.1 | - | 2 | 252 | ||
| BC132707.1 | - | 2 | 240 | 40146729 | ||
| BC132709.1 | - | 2 | 240 | 40146731 | ||