siRNA Details
| Gene Symbol: REN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000537.3 | NP_000528.1 | 8, 9 | 1095 | ||
| GenBank mRNA | BC033474.1 | - | 8, 9 | 1077 | 5188566 | |
| BC047752.1 | - | 8, 9 | 1080 | 5590211 | ||
| CR536498.1 | - | 8, 9 | 1051 | |||

REN
Genomic Map