siRNA Details
| Gene Symbol: CACNA2D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172364.4 | NP_758952.4 | 36, 37 | 3451 | ||
| XM_011521041.2 | XP_011519343.1 | 35, 36 | 7290 | |||
| GenBank mRNA | AF516695.1 | - | 36, 37 | 3583 | ||
| AK301234.1 | - | 9, 10 | 692 | |||
| AL137658.1 | - | 13, 14 | 990 | |||
| BC048288.1 | - | 8, 9 | 557 | |||
| BC150186.1 | - | 35, 36 | 3374 | 40148321 | ||
| BI908421.1 | - | 9, 10 | 659 | |||
| BX537436.1 | - | 34, 35 | 4051 | |||
| BX537437.1 | - | 36, 37 | 3452 | |||

CACNA2D4
Genomic Map