siRNA Details
| Gene Symbol: ASXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164603.1 | NP_001158075.1 | 1, 2 | 493 | ||
| NM_015338.5 | NP_056153.2 | 1, 2 | 493 | |||
| XM_006723727.3 | XP_006723790.1 | 1, 2 | 775 | |||
| XM_017027706.1 | XP_016883195.1 | 1, 2 | 775 | |||
| GenBank mRNA | AJ438952.2 | - | 1, 2 | 319 | ||
| BC064984.1 | - | 1, 2 | 478 | |||
| BC100280.1 | - | 1, 2 | 487 | |||
| BC137278.1 | - | 2, 3 | 256 | 9021275 | ||
| BC137280.1 | - | 2, 3 | 256 | 9021277 | ||
| BQ064719.1 | - | 485 | ||||

ASXL1
Genomic Map