siRNA Details
| Gene Symbol: SLC25A20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000387.5 | NP_000378.1 | 4 | 605 | ||
| GenBank mRNA | AK301744.1 | - | 5 | 566 | ||
| AK312962.1 | - | 4 | 498 | |||
| BC001689.2 | - | 4 | 477 | 3050073 | ||
| BI913588.1 | - | 4 | 508 | |||
| Y10319.1 | - | 4 | 468 | |||

SLC25A20
Genomic Map