siRNA Details
| Gene Symbol: C5orf63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164478.1 | NP_001157950.1 | 3 | 220 | ||
| NM_001164479.1 | NP_001157951.1 | 3 | 220 | |||
| GenBank mRNA | AK126569.1 | - | 3 | 208 | ||
| DA507232.1 | - | 3 | 220 | |||

C5orf63
Genomic Map