siRNA Details
| Gene Symbol: PET100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171155.1 | NP_001164626.1 | 2 | 138 | ||
| NR_033242.1 | 2 | 138 | ||||
| GenBank mRNA | AK124717.1 | - | 1 | 826 | ||
| BC150496.1 | - | 2 | 116 | |||
| CB119056.1 | - | 2 | 129 | |||
| CD641681.1 | - | 2 | 132 | |||

PET100
Genomic Map