siRNA Details
| Gene Symbol: LINC01011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055096.1 | - | 1 | 674 | ||
| AK123663.1 | - | 2 | 1010 | |||
| BC137372.1 | - | 1 | 576 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01011| Gene Symbol: LINC01011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055096.1 | - | 1 | 674 | ||
| AK123663.1 | - | 2 | 1010 | |||
| BC137372.1 | - | 1 | 576 | |||