siRNA Details
Gene Symbol: HTATSF1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033884.1 | 1 | 1606 | |||
GenBank mRNA | AK128391.1 | - | 1 | 731 | ||
BX648511.1 | - | 1 | 726 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HTATSF1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033884.1 | 1 | 1606 | |||
GenBank mRNA | AK128391.1 | - | 1 | 731 | ||
BX648511.1 | - | 1 | 726 |