siRNA Details
| Gene Symbol: LINC00997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036501.1 | 1 | 3850 | |||
| GenBank mRNA | AK092714.1 | - | 1 | 1857 | ||
| AK127989.1 | - | 1 | 1089 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00997| Gene Symbol: LINC00997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036501.1 | 1 | 3850 | |||
| GenBank mRNA | AK092714.1 | - | 1 | 1857 | ||
| AK127989.1 | - | 1 | 1089 | |||