siRNA Details
| Gene Symbol: LRRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161528.1 | NP_001155000.1 | 2 | 2120 | ||
| GenBank mRNA | BC026112.1 | - | 2 | 656 | ||
| BC144674.1 | - | 2 | 215 | 9053208 | ||
| BC144678.1 | - | 2 | 238 | 9053212 | ||

LRRD1
Genomic Map