siRNA Details
| Gene Symbol: FPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002030.4 | NP_002021.3 | 2 | 1099 | ||
| XM_011526687.2 | XP_011524989.1 | 2 | 990 | |||
| GenBank mRNA | AK123030.1 | - | 3 | 1042 | ||
| AK292541.1 | - | 3 | 1088 | |||
| BC059388.1 | - | 2 | 1082 | 30332008 | ||
| BC069070.1 | - | 1 | 903 | 7216883 | ||
| BC069593.1 | - | 1 | 903 | 7262465 | ||
| BC069812.1 | - | 1 | 903 | 7262453 | ||
| M76673.1 | - | 1 | 903 | |||
More Information
Set Membership: |
|

FPR3
Genomic Map