siRNA Details
| Gene Symbol: LINC00998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015442.1 | 2 | 114 | |||
| NR_024412.1 | 2 | 202 | ||||
| GenBank mRNA | BC065765.1 | - | 2 | 90 | ||
| BC107860.1 | - | 2 | 118 | |||
| BX537645.1 | - | 2 | 87 | |||
| DN995392.1 | - | 2 | 176 | |||

LINC00998
Genomic Map