siRNA Details
| Gene Symbol: LINC00998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015442.1 | 3 | 217 | |||
| NR_024412.1 | 3 | 305 | ||||
| GenBank mRNA | BC065765.1 | - | 3 | 193 | ||
| BC107860.1 | - | 3 | 221 | |||
| BX537645.1 | - | 3 | 190 | |||
| DN995392.1 | - | 3 | 279 | |||

LINC00998
Genomic Map