siRNA Details
| Gene Symbol: PRRT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114726.2 | NP_001108198.2 | 2 | 351 | ||
| NM_001174164.1 | NP_001167635.1 | 3 | 468 | |||
| XM_005250342.3 | XP_005250399.1 | 2 | 468 | |||
| XM_005250343.3 | XP_005250400.1 | 2 | 377 | |||
| XM_006715985.3 | XP_006716048.1 | 2 | 481 | |||
| XM_017012205.1 | XP_016867694.1 | 3 | 1287 | |||
| XM_017012206.1 | XP_016867695.1 | 2 | 1260 | |||
| GenBank mRNA | BX355277.2 | - | 3 | 414 | ||

PRRT4
Genomic Map