siRNA Details
| Gene Symbol: RAB19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008749.2 | NP_001008749.2 | 2 | 249 | ||
| XM_005249996.3 | XP_005250053.1 | 2 | 532 | |||
| XM_005249997.3 | XP_005250054.1 | 2 | 551 | |||
| XM_005249998.3 | XP_005250055.1 | 2 | 552 | |||
| XM_017012207.1 | XP_016867696.1 | 2 | 445 | |||
| GenBank mRNA | BC140796.1 | - | 1 | 119 | 9021664 | |
| BC171745.1 | - | 1 | 119 | |||
| DN996121.1 | - | 2 | 223 | |||

RAB19
Genomic Map