siRNA Details
| Gene Symbol: MFSD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152778.2 | NP_689991.1 | 13 | 2390 | ||
| XM_005262893.1 | XP_005262950.1 | 12 | 2304 | |||
| XM_005262896.1 | XP_005262953.1 | 11 | 2157 | |||
| XM_005262897.2 | XP_005262954.1 | 10 | 2103 | |||
| XM_005262898.2 | XP_005262955.1 | 10 | 2887 | |||
| XM_011531830.1 | XP_011530132.1 | 11 | 2190 | |||
| XM_011531831.1 | XP_011530133.1 | 9 | 1989 | |||
| XM_011531832.1 | XP_011530134.1 | 9 | 2773 | |||
| XM_017007989.1 | XP_016863478.1 | 8 | 2686 | |||
| GenBank mRNA | CB956202.1 | - | 1 | 396 | ||

MFSD8
Genomic Map