siRNA Details
| Gene Symbol: MAGEL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019066.4 | NP_061939.3 | 1 | 3369 | ||
| GenBank mRNA | AJ243531.1 | - | 1 | 1354 | ||
| BC035839.1 | - | 1 | 1491 | |||
| BC063834.1 | - | 1 | 1551 | |||
| BC112257.1 | - | 1 | 1291 | |||
| BC112259.1 | - | 1 | 1291 | |||

MAGEL2
Genomic Map