siRNA Details
| Gene Symbol: TOMM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001790.2 | NP_001001790.1 | 2 | 514 | ||
| NM_001134484.1 | NP_001127956.1 | 2 | 673 | |||
| NM_001134485.1 | NP_001127957.1 | 2 | 616 | |||
| GenBank mRNA | BC034247.1 | - | 2 | 446 | 3861303 | |

TOMM5
Genomic Map