siRNA Details
| Gene Symbol: DSP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008844.2 | NP_001008844.1 | 3 | 641 | ||
| NM_001319034.1 | NP_001305963.1 | 3 | 641 | |||
| NM_004415.3 | NP_004406.2 | 3 | 641 | |||
| GenBank mRNA | AB209992.1 | - | 3 | 599 | ||
| AB621818.1 | - | 3 | 546 | |||
| AK130229.1 | - | 2 | 422 | |||
| AK304182.1 | - | 3 | 544 | |||
| BC140802.1 | - | 3 | 574 | 9021670 | ||
| HM151899.1 | - | 3 | 579 | |||
| M77830.3 | - | 3 | 579 | |||

DSP
Genomic Map