siRNA Details
| Gene Symbol: ARSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000046.3 | NP_000037.2 | 8 | 2795 | ||
| XM_011543390.1 | XP_011541692.1 | 9 | 2008 | |||
| GenBank mRNA | AK314903.1 | - | 8 | 1580 | ||
| J05225.1 | - | 8 | 1515 | |||
| M32373.1 | - | 8 | 2068 | |||

ARSB
Genomic Map