siRNA Details
| Gene Symbol: ARSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000046.3 | NP_000037.2 | 4 | 2173 | ||
| NM_198709.2 | NP_942002.1 | 5 | 1242 | |||
| XM_011543390.1 | XP_011541692.1 | 5 | 1386 | |||
| XM_011543391.2 | XP_011541693.1 | 4 | 949 | |||
| XM_011543392.2 | XP_011541694.1 | 4 | 949 | |||
| XM_011543393.1 | XP_011541695.1 | 4 | 949 | |||
| XM_017009471.1 | XP_016864960.1 | 4 | 949 | |||
| GenBank mRNA | AK290865.1 | - | 4 | 949 | ||
| AK314903.1 | - | 4 | 958 | |||
| BC029051.1 | - | 5 | 1242 | 5186657 | ||
| J05225.1 | - | 4 | 893 | |||
| M32373.1 | - | 4 | 1446 | |||

ARSB
Genomic Map