siRNA Details
Gene Symbol: ARSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198709.2 | NP_942002.1 | 8 | 1969 | ||
GenBank mRNA | AK290865.1 | - | 7 | 1676 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ARSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198709.2 | NP_942002.1 | 8 | 1969 | ||
GenBank mRNA | AK290865.1 | - | 7 | 1676 |