siRNA Details
Gene Symbol: PLA2G6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK300459.1 | - | 10 | 1446 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PLA2G6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK300459.1 | - | 10 | 1446 |