siRNA Details
| Gene Symbol: PMM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000303.2 | NP_000294.1 | 2 | 175 | ||
| XM_005255374.4 | XP_005255431.1 | 2 | 175 | |||
| GenBank mRNA | AK291537.1 | - | 2 | 146 | ||
| AK296123.1 | - | 2 | 168 | |||
| AK296630.1 | - | 2 | 121 | |||
| AK299979.1 | - | 2 | 146 | |||
| AK300785.1 | - | 2 | 151 | |||
| AK304320.1 | - | 2 | 138 | |||
| AK311035.1 | - | 2 | 146 | |||
| AK312532.1 | - | 2 | 146 | |||
| BC008310.1 | - | 2 | 117 | 3611382 | ||
| DC366756.1 | - | 2 | 182 | |||
| KU178265.1 | - | 2 | 105 | |||
| KU178266.1 | - | 2 | 105 | |||
| U85773.1 | - | 2 | 153 | |||

PMM2
Genomic Map