siRNA Details
| Gene Symbol: KCNQ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004700.3 | NP_004691.2 | 8 | 1187 | ||
| NM_172163.2 | NP_751895.1 | 8 | 1187 | |||
| XM_017002792.1 | XP_016858281.1 | 5 | 428 | |||
| GenBank mRNA | AF105202.1 | - | 8 | 1187 | ||
| AK074957.1 | - | 5 | 424 | |||
| DA782056.1 | - | 5 | 479 | |||

KCNQ4
Genomic Map