siRNA Details
| Gene Symbol: C9orf152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012993.2 | NP_001013011.2 | 2 | 802 | ||
| GenBank mRNA | AK290278.1 | - | 2 | 211 | ||
| BC110807.1 | - | 2 | 193 | 5178248 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9orf152| Gene Symbol: C9orf152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012993.2 | NP_001013011.2 | 2 | 802 | ||
| GenBank mRNA | AK290278.1 | - | 2 | 211 | ||
| BC110807.1 | - | 2 | 193 | 5178248 | ||