siRNA Details
| Gene Symbol: AHI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134830.1 | NP_001128302.1 | 19 | 3166 | ||
| NM_001134831.1 | NP_001128303.1 | 21 | 3313 | |||
| NM_001134832.1 | NP_001128304.1 | 20 | 3228 | |||
| NM_017651.4 | NP_060121.3 | 20 | 3228 | |||
| XM_011535910.2 | XP_011534212.1 | 22 | 3457 | |||
| XM_011535911.2 | XP_011534213.1 | 20 | 3187 | |||
| XM_017010977.1 | XP_016866466.1 | 21 | 3432 | |||
| XM_017010978.1 | XP_016866467.1 | 20 | 3130 | |||
| XM_017010979.1 | XP_016866468.1 | 20 | 3130 | |||
| XM_017010980.1 | XP_016866469.1 | 20 | 3130 | |||
| XM_017010981.1 | XP_016866470.1 | 20 | 3265 | |||
| XM_017010982.1 | XP_016866471.1 | 20 | 3130 | |||
| XM_017010983.1 | XP_016866472.1 | 20 | 3130 | |||
| XM_017010984.1 | XP_016866473.1 | 20 | 3130 | |||
| GenBank mRNA | AJ459824.1 | - | 20 | 3208 | ||
| AJ459825.1 | - | 20 | 3208 | |||
| AJ606362.1 | - | 19 | 3146 | |||
| AK000076.1 | - | 12 | 1629 | |||
| AK024085.1 | - | 8 | 1121 | |||
| AK092262.1 | - | 19 | 3139 | |||
| AK225210.1 | - | 12 | 1629 | |||
| AL136797.1 | - | 18 | 2947 | |||
| BC015019.1 | - | 10 | 1427 | |||
| BC094800.1 | - | 21 | 3236 | 4823262 | ||
| DQ090887.1 | - | 21 | 3267 | |||

AHI1
Genomic Map