siRNA Details
| Gene Symbol: C9orf152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012993.2 | NP_001013011.2 | 2 | 1157 | ||
| GenBank mRNA | AK290278.1 | - | 2 | 566 | ||
| BC110807.1 | - | 2 | 548 | 5178248 | ||
| BX648492.1 | - | 2 | 558 | |||
| BX648620.1 | - | 2 | 577 | |||

C9orf152
Genomic Map