siRNA Details
| Gene Symbol: NDUFA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193375.1 | NP_001180304.1 | 1 | 158 | ||
| NM_175614.4 | NP_783313.1 | 1 | 158 | |||
| NR_034166.2 | 1 | 158 | ||||
| GenBank mRNA | BI160094.1 | - | 1 | 153 | ||
| BP432135.1 | - | 1 | 162 | |||

NDUFA11
Genomic Map